3,4,7,8-Tetramethyl-1,10-phenanthroline 98+%

Supplier: Thermo Scientific Chemicals

17222-0010 17222-0050
TS17222-0010EA 103 USD
TS17222-0010 TS17222-0050
3,4,7,8-Tetramethyl-1,10-phenanthroline 98+%
3,4,7,8-Tetramethyl-1,10-phenanthroline
Formula: C₁₆H₁₆N₂
MW: 236.32 g/mol
Melting Pt: 277…280 °C
Storage Temperature: Ambient
MDL Number: MFCD00004974
CAS Number: 1660-93-1
EINECS: 216-762-4

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 %
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR