3,4-(Methylenedioxy)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

17147-0100 17147-0500
TS17147-0100EA 37 USD
TS17147-0100 TS17147-0500
3,4-(Methylenedioxy)aniline 97%
3,4-(Methylenedioxy)aniline
Formula: C₇H₇NO₂
Boiling Pt: 144 °C (16 mmHg)
Melting Pt: 38…43 °C
Flash Pt: 142 °C
Storage Temperature: Ambient
MDL Number: MFCD00005832
CAS Number: 14268-66-7
EINECS: 238-161-6

Order Now

Specification Test Results

Appearance Beige to brown to black-brown Crystals and/or chunks or fused solid
Infrared spectrum Conforms
Melting point 40 to 46 °C
GC ≥96.0 %
Titration with HClO4 96.0 to 104.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR