6-Chloropurine 99+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloro-7H-purine, 6-Chloro-1H-purine, 6-Chloro-9H-purine

17128-0050 17128-0250
TS17128-0050EA 65 USD
TS17128-0050 TS17128-0250
6-Chloropurine 99+%
6-Chloropurine
Formula: C₅H₃ClN₄
MW: 154.56 g/mol
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00075825
CAS Number: 87-42-3
EINECS: 201-745-6
Merck Index: 13,02180

Order Now

Specification Test Results

Appearance Yellow to orange Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤0.5 % (105°C, 5 hrs)
Sulfated ash ≤0.1 % (800°C)
Appearance of solution (0.1% soln. in 0.1 M HCl) clear, opalescent to yellowish
Titration with HClO4 99.0 to 101.0 %
Carbon (C) 38.47 to 39.25 % (On dry substance)
Nitrogen (N) 32.52 to 36.97 % (On dry substance)
Heavy metals (as Pb) ≤10 ppm

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR