Quinaldic acid 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17105-0100
17105-0500
TS17105-0100EA
120
USD
TS17105-0100
TS17105-0500
Quinaldic acid 99%
Quinaldic acid
Formula:
C₁₀H₇NO₂ Melting Pt: 159…161 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006752 CAS Number: 93-10-7 EINECS: 202-218-3 Merck Index: 12,08227 |
Specification Test Results
Appearance | Light brown Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 157 to 159 °C |
Titration with NaOH | ≥98.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...