3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
17101-0010
17101-0050
17101-0250
TS17101-0010EA
56
USD
TS17101-0010
TS17101-0050
TS17101-0250
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate 97+%
3-(2-Pyridyl)-5,6-diphenyl-1,2,4-triazine-p,p'-disulfonic acid disodium salt hydrate
Formula:
C₂₀H₁₂N₄Na₂O₆S₂·xH₂O Melting Pt: >300 °C Storage Temperature: Ambient |
MDL Number:
MFCD00717621 CAS Number: 28048-33-1 EINECS: 248-797-6 |
Specification Test Results
Appearance | Yellow to yellow-green Powder |
Infrared spectrum | Conforms |
Water | ≤10.0 % (K.F.) |
Sulfated ash | 20.0 % to 27.8 % |
Solubility | (5 % acetate-buffer pH 5.5) Clear yellow |
UV Lambda max | 280 to 288 nm (in water) |
Specific extinction E (1%/1cm) | 385 to 465 (at lambda max) |
Titration with HClO4 | ≥97.0 % (On dried substance) |
Suitability | (for Cu + Fe determination) Passes test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...