2-Furonitrile 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Cyanofuran

17043-0050 17043-0250
TS17043-0050EA 98 USD
TS17043-0050 TS17043-0250
2-Furonitrile 99%
2-Furonitrile
Formula: C₅H₃NO
MW: 93.08 g/mol
Boiling Pt: 146 °C (1013 hPa)
Density: 1.06 g/cm³
Flash Pt: 35 °C
MDL Number: MFCD00003223
CAS Number: 617-90-3
EINECS: 210-537-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear light yellow to light orange Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4780 to 1.4800 (20°C, 589 nm)
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR