Pentafluoropyridine 99+%

Supplier: Thermo Scientific Chemicals
Danger

16999-0050 16999-0250
TS16999-0050EA 45 USD
TS16999-0050 TS16999-0250
Pentafluoropyridine 99+%
Pentafluoropyridine
Formula: C₅F₅N
Boiling Pt: 100 °C (1013 hPa)
Melting Pt: –41 °C
Density: 1.54 g/cm³
Flash Pt: 23 °C
Storage Temperature: Ambient
MDL Number: MFCD00006225
CAS Number: 700-16-3
EINECS: 211-839-9
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.3850 to 1.3870 (20°C, 589 nm)
GC ≥99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR