4-Methylquinoline 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
16977-0050
16977-0250
16977-1000
TS16977-0050EA
39
USD
TS16977-0050
TS16977-0250
TS16977-1000
4-Methylquinoline 99%
4-Methylquinoline
Formula:
C₁₀H₉N MW: 143.19 g/mol Boiling Pt: 261 °C (1013 hPa) Melting Pt: 9…10 °C Density: 1.08 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006784 CAS Number: 491-35-0 EINECS: 207-734-2 Merck Index: 13,05460 |
Specification Test Results
Appearance | Clear light yellow to green to brown Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.6180 to 1.6220 (20°C, 589 nm) |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...