2-Benzoxazolinone 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: benzo[d]oxazol-2(3H)-one

16908-0250 16908-1000 16908-5000
TS16908-0250EA 58 USD
TS16908-0250 TS16908-1000 TS16908-5000
2-Benzoxazolinone 98%
2-Benzoxazolinone
Formula: C₇H₅NO₂
MW: 135.12 g/mol
Melting Pt: 137…141 °C
Flash Pt: 160 °C
Storage Temperature: Ambient
MDL Number: MFCD00005716
CAS Number: 59-49-4
EINECS: 200-430-0
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light beige to grey-brown Powder
Infrared spectrum Conforms
Melting point 137 to 141 °C
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR