6-Chloronicotinic acid 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Chloropyridine-3-carboxylic acid, 6-Chloro-3-pyridinecarboxylic acid

16869-0050 16869-0250 16869-1000
TS16869-0050EA 44 USD
TS16869-0050 TS16869-0250 TS16869-1000
6-Chloronicotinic acid 99%
6-Chloronicotinic acid
Formula: C₆H₄ClNO₂
MW: 157.56 g/mol
Boiling Pt: 284 °C (1013 hPa)
Melting Pt: 186…190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006241
CAS Number: 5326-23-8
EINECS: 226-201-5

Order Now

Specification Test Results

Appearance Off-white to beige Crystalline powder
Infrared spectrum Conforms
Melting point 186 to 190 °C
Water ≤0.2 % (K.F.)
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR