Betahistine dihydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Methyl-2-(pyridin-2-yl)ethanamine dihydrochloride

16799-0010 167990050
TS16799-0010EA 42 USD
TS16799-0010 TS167990050
Betahistine dihydrochloride ≥98%
Betahistine dihydrochloride
Formula: C₈H₁₂N₂·2HCl
MW: 209.12 g/mol
Melting Pt: 150…154 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00012813
CAS Number: 5579-84-0

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder
Infrared spectrum Conforms
Melting point 150 to 154 °C
Loss on drying ≤1 % (1 g, 105 °C)
Solubility (5 g in 50 ml water) Clear solution
Titration with NaOH 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR