2-(2-Pyridyl)benzimidazole 99%

Supplier: Thermo Scientific Chemicals
Warning

16795-0050 16795-0100
TS16795-0050EA 76 USD
TS16795-0050 TS16795-0100
2-(2-Pyridyl)benzimidazole 99%
2-(2-Pyridyl)benzimidazole
Formula: C₁₂H₉N₃
Melting Pt: 220…225 °C
Storage Temperature: Ambient
MDL Number: MFCD00005586
CAS Number: 1137-68-4

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
Melting point 220 to 225 °C
HPLC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR