N-Carbethoxy-4-piperidone 98.5+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Piperidone-N-carboxylic acid ethyl ester, N-Ethoxycarbonyl-4-piperidone, Ethyl4-oxo-1-piperidinecarboxylate

16724-0100 16724-2500
TS16724-0100EA 32 USD
TS16724-0100 TS16724-0500 TS16724-2500
N-Carbethoxy-4-piperidone 98.5+%
N-Carbethoxy-4-piperidone
Formula: C₈H₁₃NO₃
MW: 171.19 g/mol
Boiling Pt: 95…98 °C (1 mmHg)
Density: 1.135 g/cm³
Flash Pt: 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00006188
CAS Number: 29976-53-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4740 to 1.4760 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
Density (20°C) 1.13 to 1.14 kg/l
GC ≥98.50 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR