3-Amino-4-pyrazolecarbonitrile 98%

Supplier: Thermo Scientific Chemicals
Warning

16685-0010 16685-0050 16685-0250
TS16685-0010EA 41 USD
TS16685-0010 TS16685-0050 TS16685-0250
3-Amino-4-pyrazolecarbonitrile 98%
3-Amino-4-pyrazolecarbonitrile
Formula: C₄H₄N₄
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD00005237
CAS Number: 16617-46-2
UN: 2811
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Light yellow to orange-brown Crystalline powder
Infrared spectrum Conforms
Melting point 172 to 176 °C
Titration with HClO4 ≥97.5 %
Acid value ≤2.0 mg KOH/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR