5-Bromothiophene-2-carbaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Bromo-2-thiophenecarboxaldehyde

16665-0050 16665-0250 16665-1000
TS16665-0050EA 45 USD
TS16665-0050 TS16665-0250 TS16665-1000
5-Bromothiophene-2-carbaldehyde 97%
5-Bromothiophene-2-carbaldehyde
Formula: C₅H₃BrOS
MW: 191.05 g/mol
Density: 1.61 g/cm³
Flash Pt: 98 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00005432
CAS Number: 4701-17-1

Order Now

Specification Test Results

Appearance Clear yellow to brown Liquid after melting
Infrared spectrum Conforms
Refractive index 1.6380 to 1.6410 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR