1-Isoquinolinol 98%

Supplier: Thermo Scientific Chemicals

16646-0010 16646-0050
TS16646-0010EA 72 USD
TS16646-0010 TS16646-0050
1-Isoquinolinol 98%
1-Isoquinolinol
Formula: C₉H₇NO
Melting Pt: 211…214 °C
Storage Temperature: Ambient
MDL Number: MFCD00006899
CAS Number: 491-30-5

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 210 to 215 °C
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR