N-Ethylcarbazole-3-carboxaldehyde 94% tech.

Supplier: Thermo Scientific Chemicals

Synonyms: 3-Formyl-N-ethylcarbazole

16603-0250 16603-1000 166035000
TS16603-0250EA 105 USD
TS16603-0250 TS16603-1000 TS166035000
N-Ethylcarbazole-3-carboxaldehyde 94% tech.
N-Ethylcarbazole-3-carboxaldehyde
Formula: C₁₅H₁₃NO
MW: 223.28 g/mol
Melting Pt: 85…91 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00004963
CAS Number: 7570-45-8

Order Now

Specification Test Results

Appearance Yellow to ochre Crystalline powder or granules
Infrared spectrum Conforms
Melting point 85 to 91 °C
HPLC ≥92 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR