1-(2-Pyridyl)piperazine 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-(1-Piperazinyl)pyridine

16590-0050 16590-0250 16590-1000
TS16590-0050EA 51 USD
TS16590-0050 TS16590-0250 TS16590-1000
1-(2-Pyridyl)piperazine 98%
1-(2-Pyridyl)piperazine
Formula: C₉H₁₃N₃
Boiling Pt: 120…122 °C (2 mmHg)
Density: 1.07 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00006216
CAS Number: 34803-66-2
EINECS: 252-220-3

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5930 to 1.5960 (20°C, 589 nm)
GC ≥97.5 %
Sulfate (SO4) ≤0.01 %
Arsenic (As) ≤3 ppm
Lead (Pb) ≤0.002 %
Insoluble matter ≤0.005 %
Alkalies + earths ≤0.2 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR