5-Bromocytosine 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Amino-5-bromo-2-pyrimidinol, 4-Amino-5-bromopyrimidin-2(1H)-one, 4-Amino-5-bromo-2-hydroxypyrimidine

16578-0010 16578-2500
TS16578-0010EA 149 USD
TS16578-0010 TS16578-2500
5-Bromocytosine 99%
5-Bromocytosine
Formula: C₄H₄BrN₃O
MW: 190 g/mol
Melting Pt: 240…243 °C
MDL Number: MFCD00056025
CAS Number: 2240-25-7
EINECS: 218-806-8

Order Now

Specification Test Results

Appearance Light yellow Powder
Infrared spectrum Conforms
Assay ≥98.5 %
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR