4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Hexahydro-4-methylphthalic anhydride

16539-0025 16539-5000
TS16539-0025EA 464 USD
TS16539-0025 TS16539-5000
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres) 98%
4-Methyl-1,2-cyclohexanedicarboxylic anhydride (mixture of cis- and trans-isomeres)
Formula: C₉H₁₂O₃
Boiling Pt: 120 °C (1mbar)
Melting Pt: –29 °C
Density: 1.16 g/cm³
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00005927
CAS Number: 19438-60-9

Order Now

Specification Test Results

Appearance Clear liquid
Infrared spectrum Conforms
Refractive index 1.4764 to 1.4784 (20°C, 589 nm)
GC ≥97.5 %
Color scale ≤100 APHA


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR