4-Phenylpiperidine 98%

Supplier: Thermo Scientific Chemicals
Warning

16477-0010 16477-0050
TS16477-0010EA 47 USD
TS16477-0010 TS16477-0050
4-Phenylpiperidine 98%
4-Phenylpiperidine
Formula: C₁₁H₁₅N
MW: 161.25 g/mol
Boiling Pt: 255…257 °C (727 mmHg)
Melting Pt: 57…63 °C
Storage Temperature: Ambient
MDL Number: MFCD00006002
CAS Number: 771-99-3
EINECS: 212-243-1

Order Now

Specification Test Results

Appearance Off-white to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 59 to 64 °C
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR