4-Pyridinethiol 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Pyridine-4-thiol, 4-Mercaptopyridine

16471-0010 16471-0100
TS16471-0010EA 45 USD
TS16471-0010 TS16471-0100
4-Pyridinethiol 96%
4-Pyridinethiol
Formula: C₅H₅NS
MW: 111.17 g/mol
Melting Pt: 182…189 °C
Storage Temperature: Freezer
MDL Number: MFCD00006422
CAS Number: 4556-23-4
EINECS: 224-926-1

Order Now

Specification Test Results

Appearance Yellow to yellow-green to brown Crystalline powder
Infrared spectrum Conforms
Titration with NaOH ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR