Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl 4-chloro-2-methylthio-5-pyrimidinecarboxylate, Ethyl 4-chloro-2-(methylsulphanyl)pyrimidine-5-carboxylate, Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
16375-0250
16375-1000
TS16375-0250EA
164
USD
TS16375-0250
TS16375-1000
Ethyl-4-chloro-2-(methylthio)pyrimidine-5-carboxylate 98%
Ethyl 4-chloro-2-(methylthio)pyrimidine-5-carboxylate
Formula:
C₈H₉ClN₂O₂S MW: 232.69 g/mol Melting Pt: 60…63 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006085 CAS Number: 5909-24-0 |
Specification Test Results
Appearance | White to yellow Powder |
Infrared spectrum | Conforms |
Melting point | 59 to 64 °C |
GC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...