3-Methoxy-2(1H)-pyridone 97%

Supplier: Thermo Scientific Chemicals
Warning

16354-0010 16354-0050
TS16354-0010EA 88 USD
TS16354-0010 TS16354-0050
3-Methoxy-2(1H)-pyridone 97%
3-Methoxy-2(1H)-pyridone
Formula: C₆H₇NO₂
MW: 125.1253 g/mol
Melting Pt: 113…115 °C
Storage Temperature: Ambient
MDL Number: MFCD00006015
CAS Number: 20928-63-6

Order Now

Specification Test Results

Appearance White to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR