δ-Valerolactone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pentan-5-olide, Tetrahydro-2H-pyran-2-one, 5-Hydroxypentanoic acid lactone, δ-Valerolactone

16271-0010 16271-0500 16271-2500
TS16271-0010EA 933 USD
TS16271-0010 TS16271-0500 TS16271-2500
δ-Valerolactone 99%
δ-Valerolactone
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 230 °C (760 mmHg)
Melting Pt: –13 °C
Density: 1.1 g/cm³
Flash Pt: 112 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006645
CAS Number: 542-28-9
EINECS: 208-807-1

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4560 to 1.4590 (20°C, 589 nm)
Additional info remark: may contain variable amounts of polymerized product
GC ≥98.5 %
pH 4.5 to 6.5 (4 % in water)
Total volatile ≤5 % (including water)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR