2,4,6-Collidine 99%

Supplier: Thermo Scientific Chemicals
Danger

16195-0010 16195-0025 16195-1000 16195-5000
TS16195-0010EA 438 USD
TS16195-0010 TS16195-0025 TS16195-1000 TS16195-5000
2,4,6-Collidine 99%
2,4,6-Collidine
Formula: C₈H₁₁N
MW: 121.18 g/mol
Boiling Pt: 171
Melting Pt: -43
Density: 0.91 g/cm³
Flash Pt: 57 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006338
CAS Number: 108-75-8
EINECS: 203-613-3
UN: 1992
ADR: 3 III
Merck Index: 13,09789

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4970 to 1.4990 (20°C, 589 nm)
GC ≥98.5 %
Sensitivity (to chlorine) Passes test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR