4-tert-Butylpyridine ≥97.5% (by GC)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-(tert-Butyl)pyridine

16194-0250 161941000
TS16194-0250EA 166 USD
TS16194-0250 TS161941000
4-tert-Butylpyridine ≥97.5% (by GC)
4-tert-Butylpyridine
Formula: C₉H₁₃N
MW: 135.21
Boiling Pt: 196
Melting Pt: -0.9
Density: 0.9219
Flash Pt: 63 °C
MDL Number: MFCD00006435
CAS Number: 3978-81-2
EINECS: 223-614-2

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4940 to 1.4960 (20°C, 589 nm)
GC ≥97.5 %
Potassium fluosilicate ≤0.1 % (K2SiF6)
Iron (Fe) ≤0.001 %
Sodium (Na) ≤0.2 %
Heavy metals (as Pb) ≤0.001 %
Titrable acid ≤0.03 meq/g
Titrable base ≤0.01 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR