4-Benzylpiperidine 98%

Supplier: Thermo Scientific Chemicals

16193-0050 16193-1000
TS16193-0050EA 17 USD
TS16193-0050 TS16193-1000
4-Benzylpiperidine 98%
4-Benzylpiperidine
Formula: C₁₂H₁₇N
Boiling Pt: 279 °C (1013 hPa)
Melting Pt: 6…7 °C
Density: 0.99 g/cm³
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00006006
CAS Number: 31252-42-3
EINECS: 250-535-0

Order Now

Specification Test Results

Appearance Clear colorless to yellow Viscous liquid
Infrared spectrum Conforms
Refractive index 1.5370 to 1.5390 (20°C, 589 nm)
GC ≥97.5 %
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR