4-Nitroimidazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4-Nitro-1H-imidazole, 5-Nitroimidazole, 5-Nitro-1H-imidazole

16160-0250 16160-1000 16160-5000
TS16160-0250EA 40 USD
TS16160-0250 TS16160-1000 TS16160-5000
4-Nitroimidazole 98%
4-Nitroimidazole
Formula: C₃H₃N₃O₂
MW: 113.08 g/mol
Melting Pt: 303 °C (dec.)
Flash Pt: >200 °C
Storage Temperature: Ambient
MDL Number: MFCD00005196
CAS Number: 3034-38-6
EINECS: 221-224-7

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR