Bathocuproin 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2,9-Dimethyl-4,7-diphenyl-1,10-phenanthroline

16134-0010 16134-0050
TS16134-0010EA 94 USD
TS16134-0010 TS16134-0050
Bathocuproin 98%
Bathocuproin
Formula: C₂₆H₂₀N₂
MW: 360.46 g/mol
Melting Pt: 279…283 °C
Storage Temperature: Ambient
MDL Number: MFCD00004972
CAS Number: 4733-39-5
EINECS: 225-240-5

Order Now

Specification Test Results

Appearance White to cream to yellow Powder
Infrared spectrum Conforms
Melting point 277 to 285 °C (Dec)
Titration with HClO4 97.5 to 102.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR