2-Methyl-4(5)-nitroimidazole 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Methyl-4-nitro-1H-imidazole, 2-Methyl-5-nitroimidazole, 2-Methyl-5-nitro-1H-imidazole, 2-Methyl-4-nitroimidazole

15947-0500 15947-2500
TS15947-0500EA 26 USD
TS15947-0500 TS15947-2500
2-Methyl-4(5)-nitroimidazole 99%
2-Methyl-4(5)-nitroimidazole
Formula: C₄H₅N₃O
Melting Pt: 252…254 °C
MDL Number: MFCD00005191
CAS Number: 696-23-1
EINECS: 211-790-3

Order Now

Specification Test Results

Appearance White to beige Powder
Infrared spectrum Conforms
Titration with HClO4 98.0 to 102.0 % (On dried substance)
Sodium dihydrogenphosphate ≤2.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR