1,2-Dimethylimidazole 96+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Dimethyl-1H-imidazole

15944-1000 15944-5000
TS15944-1000EA 39 USD
TS15944-1000 TS15944-5000
1,2-Dimethylimidazole 96+%
1,2-Dimethylimidazole
Formula: C₅H₈N₂
MW: 96.13 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: 37…39 °C
Density: 1.084 g/cm³
Flash Pt: 93 °C
Storage Temperature: Ambient
MDL Number: MFCD00005294
CAS Number: 1739-84-0
EINECS: 217-101-2

Order Now

Specification Test Results

Appearance White to yellow to orange Crystals or crystalline mass or liquid (May darken on storage)
Infrared spectrum Conforms
GC ≥96.0 %
Insoluble matter ≤0.05 % (In methanol)
Residue after reduction ≤0.02 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR