Isoquinoline-5-amine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Isoquinolin-5-amine, 5-Aminoisoquinoline
15942-0010
15942-0050
TS15942-0010EA
43
USD
TS15942-0010
TS15942-0050
Isoquinoline-5-amine 99%
Isoquinoline-5-amine
Formula:
C₉H₈N₂ MW: 144.18 g/mol Melting Pt: 127…130 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006907 CAS Number: 1125-60-6 EINECS: 214-408-3 UN: 2811 ADR: 6.1,III |
Specification Test Results
Appearance | Beige to yellow-brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 126 to 130 °C |
GC | ≥98.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...