Succinic anhydride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dihydrofuran-2,5-dione

15876-0010 15876-0050 15876-0051 15876-0500 15876-2500
TS15876-0010EA 128 USD
TS15876-0010 TS15876-0050 TS15876-0051 TS15876-0500 TS15876-2500
Succinic anhydride 99%
Succinic anhydride
Formula: C₄H₄O₃
MW: 100.07 g/mol
Boiling Pt: 261
Melting Pt: 120
Flash Pt: 157
Storage Temperature: Ambient
MDL Number: MFCD00005525
CAS Number: 108-30-5
EINECS: 203-570-0
UN: 3261
ADR: 8 II
Merck Index: 12,09039

Order Now

Specification Test Results

Appearance White Powder or crystals or flakes and/or chips
Infrared spectrum Conforms
Melting point 116 to 120 °C
Titration by coupling ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR