2-Vinylpyridine 97% stabilized

Supplier: Thermo Scientific Chemicals
Danger

15800-0010 15800-0050 15800-2500
TS15800-0010EA 238 USD
TS15800-0010 TS15800-0050 TS15800-2500
2-Vinylpyridine 97% stabilized
2-Vinylpyridine
Formula: C₇H₇N
MW: 105.14 g/mol
Boiling Pt: 159 °C (760 mmHg)
Melting Pt: –50 °C
Density: 0.9983 g/cm³
Flash Pt: 47 °C
Storage Temperature: Freezer
MDL Number: MFCD00006355
CAS Number: 100-69-6
EINECS: 202-879-8
UN: 3073
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Clear brown Liquid
Infrared spectrum Conforms
Refractive index 1.5490 to 1.5510 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer 0.1 % tert.-butylcatechol
UV at 400 nm A: ≤0.01
Color scale ≤15 APHA
Residue after evaporation ≤0.01 %
Heavy metals (as Pb) ≤0.001 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR