Thymine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Methylpyrimidine-2,4(1H,3H)-dione, 5-Methyluracil

15785-0050 15785-0250 15785-1000 15785-5000
TS15785-0050EA 23 USD
TS15785-0050 TS15785-0250 TS15785-1000 TS15785-5000
Thymine 99%
Thymine
Formula: C₅H₆N₂O₂
MW: 126.11 g/mol
Boiling Pt: 389 °C (1013 hPa)
Melting Pt: 316…317 °C
Storage Temperature: Ambient
MDL Number: MFCD00006026
CAS Number: 65-71-4
EINECS: 200-616-1
Merck Index: 12,09539

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
HPLC ≥98.5 %
Chloride (Cl) ≤0.005 %
Sulfate (SO4) ≤0.01 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.002 %
Sodium (Na) ≤0.005 %
Insoluble matter ≤0.005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR