(±)-Tetrahydrofurfurylamine 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Tetrahydrofurfurylamine

15782-0050 15782-1000 15782-5000
TS15782-0050EA 21 USD
TS15782-0050 TS15782-1000 TS15782-5000
(±)-Tetrahydrofurfurylamine 97%
(±)-Tetrahydrofurfurylamine
Formula: C₅H₁₁NO
MW: 101.15 g/mol
Boiling Pt: 153…154 °C (744 mmHg)
Density: 0.98 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00005373
CAS Number: 4795-29-3
EINECS: 225-351-9
UN: 2943
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to dark yellow Liquid (May darken on storage)
Infrared spectrum Conforms
Refractive index 1.4540 to 1.4570 (20°C, 589 nm)
Titration with HClO4 ≥96.0 %
UV at 400 nm A: ≤0.01
Color scale ≤10 APHA
Residue after evaporation ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR