Pyrimidine 99%

Supplier: Thermo Scientific Chemicals
Warning

15770-0050 15770-0250
TS15770-0050EA 105 USD
TS15770-0050 TS15770-0250
Pyrimidine 99%
Pyrimidine
Formula: C₄H₄N₂
Boiling Pt: 123…124 °C (760 mmHg)
Melting Pt: 20…22 °C
Density: 1.01 g/cm³
Flash Pt: 31 °C
Storage Temperature: Ambient
MDL Number: MFCD00006059
CAS Number: 289-95-2
EINECS: 206-026-0
UN: 1993
ADR: 3,III
Merck Index: 12,08170

Order Now

Specification Test Results

Appearance Clear colorless to orange Liquid or solid
Infrared spectrum Conforms
Refractive index 1.5010 to 1.5050 (20°C, 589 nm)
GC ≥98.5 %
Phosphate (PO4) ≤0.0005 %
Sulfate (SO4) ≤0.005 %
Ammonium (NH4) ≤0.003 %
Barium (Ba) ≤0.005 %
Calcium (Ca) ≤0.01 %
Iron (Fe) ≤0.0005 %
Manganese (Mn) ≤0.0005 %
Potassium (K) ≤0.005 %
Sodium (Na) ≤0.005 %
Strontium (Sr) ≤0.005 %
Heavy metals (as Pb) ≤5 ppm
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR