4-Picoline 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 4-Methylpyridine, γ-Picoline

15760-0010 15760-0025 15760-2500
TS15760-0010EA 91 USD
TS15760-0010 TS15760-0025 TS15760-2500
4-Picoline 99%
4-Picoline
Formula: C₆H₇N
MW: 93.13 g/mol
Boiling Pt: 144.9
Melting Pt: 3
Density: 0.95 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00006440
CAS Number: 108-89-4
EINECS: 203-626-4
UN: 2313
ADR: 3 III
Merck Index: 12,07556

Order Now

Specification Test Results

Appearance Clear light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5040 to 1.5060 (20°C, 589 nm)
Water ≤0.2 %
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR