1-Methyl-4-piperidone 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: N-Methyl-4-piperidone

15692-1000 15692-5000
TS15692-1000EA 80 USD
TS15692-1000 TS15692-5000
1-Methyl-4-piperidone 98%
1-Methyl-4-piperidone
Formula: C₆H₁₁NO
Boiling Pt: 55…58 °C (10 mmHg)
Density: 0.98 g/cm³
Flash Pt: 58 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006191
CAS Number: 1445-73-4
EINECS: 215-895-5
UN: 2924
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear yellow to orange to brown Liquid
Infrared spectrum Conforms
Refractive index 1.4590 to 1.4630 (20°C, 589 nm)
Water ≤0.5 % (K.F.)
GC ≥97.5 %
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤0.0005 %
Magnesium (Mg) ≤0.002 %
Potassium (K) ≤0.01 %
Heavy metals (as Pb) ≤0.001 %
Insoluble matter ≤0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR