2-Mercaptopyrimidine 98%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Thiopyrimidine

15674-0100 15674-0500
TS15674-0100EA 44 USD
TS15674-0100 TS15674-0500
2-Mercaptopyrimidine 98%
2-Mercaptopyrimidine
Formula: C₄H₄N₂S
MW: 112.16 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00006073
CAS Number: 1450-85-7
EINECS: 215-917-3

Order Now

Specification Test Results

Appearance Yellow Powder
Infrared spectrum Conforms
Titration with NaOH ≥97.5 %
Phosphate (PO4) ≤5 ppm
Ammonium (NH4) ≤5 ppm
Calcium (Ca) ≤0.002 %
Iron (Fe) ≤5 ppm
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.03 %
Heavy metals (as Pb) ≤5 ppm
Sulfur compounds (as SO4) ≤0.003 %
Insoluble matter ≤0.01 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR