Homophthalic anhydride 98%

Supplier: Thermo Scientific Chemicals
Warning

15630-0050 15630-0250 15630-1000
TS15630-0050EA 37 USD
TS15630-0050 TS15630-0250 TS15630-1000
Homophthalic anhydride 98%
Homophthalic anhydride
Formula: C₉H₆O₃
Melting Pt: 140…144 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006894
CAS Number: 703-59-3
EINECS: 211-873-4

Order Now

Specification Test Results

Appearance Light yellow to beige Powder
Infrared spectrum Conforms
Titration excess NaOH ≥97.5 %
Phosphate (PO4) ≤0.003 %
Insoluble matter ≤0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR