2-Amino-5-mercapto-1,3,4-thiadiazole 98%
Supplier: Thermo Scientific Chemicals
15597-0250
15597-0500
15597-2500
TS15597-0250EA
40
USD
TS15597-0250
TS15597-0500
TS15597-2500
2-Amino-5-mercapto-1,3,4-thiadiazole 98%
2-Amino-5-mercapto-1,3,4-thiadiazole
Formula:
C₂H₃N₃S₂ Melting Pt: 231…236 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003108 CAS Number: 2349-67-9 EINECS: 219-078-4 |
Specification Test Results
Appearance | White to cream to light yellow Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤1 % (110°C) |
HPLC | ≥97.5 % |
Titration with NaOH | ≥97.5 % (On dry substance) |
UV | at 400 nm A: ≤0.03 |
Color scale | ≤10 APHA |
Residue after evaporation | ≤0.0005 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...