2,4-Dihydroxypyrimidine-5-carboxylic acid 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Isoorotic acid, Uracil-5-carboxylic acid

15520-0050 15520-0250
TS15520-0050EA 51 USD
TS15520-0050 TS15520-0250
2,4-Dihydroxypyrimidine-5-carboxylic acid 98%
2,4-Dihydroxypyrimidine-5-carboxylic acid
Formula: C₅H₄N₂O₄
MW: 156.10 g/mol
Melting Pt: 283 °C
Storage Temperature: Ambient
MDL Number: MFCD00006023
CAS Number: 23945-44-0

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration with NaOH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR