2,3-Diaminopyridine 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Pyridine-2,3-diyldiamine

15502-0050 15502-0100 15502-0250 155021000
TS15502-0050EA 69 USD
TS15502-0050 TS15502-0100 TS15502-0250 TS155021000
2,3-Diaminopyridine 98%
2,3-Diaminopyridine
Formula: C₅H₇N₃
MW: 109.13 g/mol
Boiling Pt: 148…150 °C (5 mmHg)
Melting Pt: 114…116 °C
Flash Pt: 205 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00006319
CAS Number: 452-58-4
EINECS: 207-200-9
UN: 2811
ADR: 6.1,II

Order Now

Specification Test Results

Appearance Grey to dark brown Crystalline powder or needles (May darken on storage)
Infrared spectrum Conforms
GC ≥97.5 %
Titration with HClO4 97.5 to 102.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR