Phthalic anhydride 99%

Supplier: Thermo Scientific Chemicals
Danger

15496-0010 15496-0025 15496-0100 15496-2500
TS15496-0010EA 48 USD
TS15496-0010 TS15496-0025 TS15496-0100 TS15496-2500
Phthalic anhydride 99%
Phthalic anhydride
Formula: C₈H₄O₃
MW: 148.12 g/mol
Boiling Pt: 284 °C (760 mmHg)
Melting Pt: 129…132 °C
Flash Pt: 152 °C
Storage Temperature: Ambient
MDL Number: MFCD00005918
CAS Number: 85-44-9
EINECS: 201-607-5
UN: 2214
ADR: 8,III
Merck Index: 13,07457

Order Now

Specification Test Results

Appearance White Flakes
Infrared spectrum Conforms
Melting point 129 to 133 °C
Titration by coupling ≥98.5 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR