2-Coumaranone 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Benzofuran-2(3H)-one, Benzo[b]furan-2(3H)-one

15446-0010 15446-0050 15446-0100
TS15446-0010EA 16 USD
TS15446-0010 TS15446-0050 TS15446-0100
2-Coumaranone 97%
2-Coumaranone
Formula: C₈H₆O₂
MW: 134.13 g/mol
Melting Pt: 47…52 °C
Flash Pt: >112 °C
Storage Temperature: Ambient
MDL Number: MFCD00005856
CAS Number: 553-86-6
EINECS: 209-052-0

Order Now

Specification Test Results

Appearance White to yellow to light orange Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 47 to 52 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR