1-Benzyl-2-pyrrolidinone 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
1-Benzylpyrrolidin-2-one
15412-0250
15412-1000
TS15412-0250EA
27
USD
TS15412-0250
TS15412-1000
1-Benzyl-2-pyrrolidinone 97%
1-Benzyl-2-pyrrolidinone
Formula:
C₁₁H₁₃NO MW: 175.23 g/mol Density: 1.09 g/cm³ Flash Pt: >112 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003195 CAS Number: 5291-77-0 EINECS: 226-131-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.5500 to 1.5530 (20°C, 589 nm) |
GC | ≥96.0 % |
NMR (1H-NMR) | Consistent with structure |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...