cis-1,2-Cyclohexanedicarboxylic anhydride 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
15403-0010
15403-0025
15403-2500
TS15403-0010EA
133
USD
TS15403-0010
TS15403-0025
TS15403-2500
cis-1,2-Cyclohexanedicarboxylic anhydride 99%
cis-1,2-Cyclohexanedicarboxylic anhydride
Formula:
C₈H₁₀O₃ Boiling Pt: 296 °C (760 mmHg) Melting Pt: 29…32 °C Flash Pt: 143 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005926 CAS Number: 13149-00-3 EINECS: 236-086-3 |
Specification Test Results
Appearance | White to off-white Low melting crystalline solid |
Infrared spectrum | Conforms |
Melting point | 29 to 32 °C |
GC | ≥98.5 % |
Color of molten substance | ≤50 APHA |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...