2-Amino-4-methylpyridine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aminopicoline, 2-Amino-4-picoline, 4-Methyl-2-pyridylamine, 4-Methylpyridin-2-amine
15386-1000
15386-5000
TS15386-1000EA
39
USD
TS15386-1000
TS15386-5000
2-Amino-4-methylpyridine 98%
2-Amino-4-methylpyridine
Formula:
C₆H₈N₂ MW: 108.14 g/mol Boiling Pt: 230.9 Melting Pt: 98 Flash Pt: 118 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006321 CAS Number: 695-34-1 EINECS: 211-780-9 UN: 2811 ADR: 6.1 III Merck Index: 13,00465 |
Specification Test Results
Appearance | Light yellow to beige to brown Flakes |
Infrared spectrum | Conforms |
Melting point | 96 to 101 °C |
Water | ≤0.5 % |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...